Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008764536.1 | XP_008762758.1 | 2 | 7640 | |
XM_008764537.1 | XP_008762759.1 | 2 | 190 | ||
XM_008764538.1 | XP_008762760.1 | 2 | 180 | ||
XM_008764539.1 | XP_008762761.1 | 2 | 7640 |