Gene Transcripts

Gene Symbol
Tmprss12
Entrez Gene ID
100362912
Gene Name
transmembrane (C-terminal) protease, serine 12
Gene Aliases
RGD1560236
Chromosome Location
Chr. 7: 141953340 - 141966814 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765860.1 XP_008764082.1 3, 4 1074

View Full Product Details