Gene Transcripts

Gene Symbol
LOC367830
Entrez Gene ID
367830
Gene Name
similar to FAM48A protein
Gene Aliases
Chromosome Location
Chr. X: 63064950 - 63115272 on Build Rnor_6.0
UniGene ID
Rn.141105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037211.1 NP_001032288.1 2 1114
GenBank mRNA BC100149.1 - 2 1114

View Full Product Details