Gene Transcripts

Gene Symbol
Olr10
Entrez Gene ID
293072
Gene Name
olfactory receptor 10
Gene Aliases
Chromosome Location
Chr. X: 95344825 - 95345772 on Build Rnor_6.0
UniGene ID
Rn.141956
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000115.1 NP_001000115.1 1 681

View Full Product Details