Gene Transcripts

Gene Symbol
Ovol2
Entrez Gene ID
296201
Gene Name
ovo-like zinc finger 2
Gene Aliases
Zfp339
Chromosome Location
Chr. 3: 138433990 - 138463690 on Build Rnor_6.0
UniGene ID
Rn.104639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106519.1 NP_001099989.1 4 683
XM_006235124.2 XP_006235186.1 4 1842

View Full Product Details