Gene Transcripts

Gene Symbol
Sat2
Entrez Gene ID
360547
Gene Name
spermidine/spermine N1-acetyltransferase family member 2
Gene Aliases
Chromosome Location
Chr. 10: 56227513 - 56229796 on Build Rnor_6.0
UniGene ID
Rn.23795
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108278.1 NP_001101748.1 1, 2 217
XM_006246743.1 XP_006246805.1 2, 3 338
XM_006246744.2 XP_006246806.1 2, 3 239
XM_008767837.1 XP_008766059.1 2, 3 338
GenBank mRNA FQ218736.1 - 2, 3 141

View Full Product Details