Gene Transcripts

Gene Symbol
Trh
Entrez Gene ID
25569
Gene Name
thyrotropin releasing hormone
Gene Aliases
Pro-TRH, THR, TRH01, Trf
Chromosome Location
Chr. 4: 124110716 - 124113242 on Build Rnor_6.0
UniGene ID
Rn.22
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013046.3 NP_037178.1 3 720
XM_006236899.2 XP_006236961.1 3 1151
GenBank mRNA BC161830.1 - 3 667
FQ220299.1 - 1 268
FQ221570.1 - 3 696
FQ221573.1 - 3 696
FQ222015.1 - 3 696
FQ222377.1 - 3 695
FQ222571.1 - 3 694
FQ222984.1 - 3 694
FQ223091.1 - 3 696
FQ228499.1 - 3 696
FQ228758.1 - 3 696
FQ229082.1 - 3 696
FQ229292.1 - 3 694
M12138.1 - 3 687
M36317.1 - 3 687

View Full Product Details