Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001106890.2 | NP_001100360.1 | 3 | 310 | |
NM_001271131.1 | NP_001258060.1 | 3 | 266 | ||
XM_006244512.2 | XP_006244574.1 | 3 | 500 | ||
XM_008766849.1 | XP_008765071.1 | 4 | 433 | ||
XM_008766850.1 | XP_008765072.1 | 3 | 323 | ||
GenBank mRNA | FM100935.1 | - | 3 | 240 |