Gene Transcripts

Gene Symbol
Reg3g
Entrez Gene ID
24620
Gene Name
regenerating islet-derived 3 gamma
Gene Aliases
PAPIII, Pap3
Chromosome Location
Chr. 4: 109529679 - 109532372 on Build Rnor_6.0
UniGene ID
Rn.9729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173097.1 NP_775120.1 5, 6 490
XM_008763019.1 XP_008761241.1 5, 6 628
GenBank mRNA L20869.1 - 5, 6 490

View Full Product Details