Gene Transcripts

Gene Symbol
Ppy
Entrez Gene ID
24677
Gene Name
pancreatic polypeptide
Gene Aliases
Chromosome Location
Chr. 10: 90041582 - 90042877 on Build Rnor_6.0
UniGene ID
Rn.9755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012626.2 NP_036758.1 3 265
GenBank mRNA BP484139.1 - 3 248
M13588.1 - 3 265

View Full Product Details