Gene Transcripts

Gene Symbol
Serpina3c
Entrez Gene ID
24794
Gene Name
serine (or cysteine) proteinase inhibitor, clade A, member 3C
Gene Aliases
CPi-21, GHR-P63, Kbp, Klkbp, SPI-2, SPI-2.3, SPI2, Serpina3k, Spin2, Spin2b
Chromosome Location
Chr. 6: 127996162 - 128003435 on Build Rnor_6.0
UniGene ID
Rn.91257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012657.2 NP_036789.2 5 1304
XM_006240458.2 XP_006240520.1 5 1347
GenBank mRNA D00751.1 - 5 1257
FQ209577.1 - 5 1306
FQ209663.1 - 4 1032
FQ210286.1 - 5 1310
FQ218085.1 - 5 1309
FQ218647.1 - 4 981
FQ218802.1 - 5 1306
FQ219296.1 - 5 1310
FQ219312.1 - 5 1308
FQ219600.1 - 5 1304
M14320.1 - 4 802
M15916.1 - 5 1269
X05348.1 - 5 1291
X16358.1 - 5 1305

View Full Product Details