Gene Transcripts

Gene Symbol
Rell1
Entrez Gene ID
289635
Gene Name
RELT-like 1
Gene Aliases
RGD1307468
Chromosome Location
Chr. 14: 45982216 - 46041907 on Build Rnor_6.0
UniGene ID
Rn.165637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113776.1 NP_001107248.1 5 673
XM_006250998.2 XP_006251060.1 5 768
GenBank mRNA BC158872.1 - 5 673
FQ224412.1 - 5 600

View Full Product Details