Gene Transcripts

Gene Symbol
Tmco6
Entrez Gene ID
291661
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
RGD1309263
Chromosome Location
Chr. 18: 29579145 - 29585740 on Build Rnor_6.0
UniGene ID
Rn.144763
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106154.1 NP_001099624.1 10 1273
GenBank mRNA BC166835.1 - 10 1135

View Full Product Details