Gene Transcripts

Gene Symbol
Prdm6
Entrez Gene ID
307305
Gene Name
PR domain containing 6
Gene Aliases
RGD1559594
Chromosome Location
Chr. 18: 48433747 - 48538834 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772189.1 XP_008770411.1 5 1177
XM_008772190.1 XP_008770412.1 4 1451
XM_008772191.1 XP_008770413.1 5 1177

View Full Product Details