Gene Transcripts

Gene Symbol
Trappc10
Entrez Gene ID
309678
Gene Name
trafficking protein particle complex 10
Gene Aliases
Tmem1
Chromosome Location
Chr. 20: 11168298 - 11228634 on Build Rnor_6.0
UniGene ID
Rn.46413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173528.1 NP_001166999.1 21 3349

View Full Product Details