Gene Transcripts

Gene Symbol
Phf20l1
Entrez Gene ID
314964
Gene Name
PHD finger protein 20-like 1
Gene Aliases
Chromosome Location
Chr. 7: 107406948 - 107445219 on Build Rnor_6.0
UniGene ID
Rn.22288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271439.1 NP_001258368.1 1640
GenBank mRNA FQ232787.1 - 524

View Full Product Details