Gene Transcripts

Gene Symbol
Pls1
Entrez Gene ID
315926
Gene Name
plastin 1
Gene Aliases
Chromosome Location
Chr. 8: 103557361 - 103659625 on Build Rnor_6.0
UniGene ID
Rn.224870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108178.2 NP_001101648.2 6 619
XM_006243574.2 XP_006243636.1 10 1222
XM_006243575.2 XP_006243637.1 10 1602
XM_006243576.2 XP_006243638.1 10 1292
XM_008766518.1 XP_008764740.1 9 1236
XM_008766519.1 XP_008764741.1 10 1142
GenBank mRNA BC168847.1 - 6 619

View Full Product Details