Gene Transcripts

Gene Symbol
Sh2d5
Entrez Gene ID
366489
Gene Name
SH2 domain containing 5
Gene Aliases
RGD1305952
Chromosome Location
Chr. 5: 156615889 - 156624962 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239282.2 XP_006239344.1 6 1016
XM_006239283.2 XP_006239345.1 6 674
XM_006239286.2 XP_006239348.1 5 517
XM_008764300.1 XP_008762522.1 7 1004
XM_008764301.1 XP_008762523.1 7 1007
XM_008764302.1 XP_008762524.1 5 504

View Full Product Details