Gene Transcripts

Gene Symbol
Sspo
Entrez Gene ID
474348
Gene Name
SCO-spondin
Gene Aliases
Scospondin
Chromosome Location
Chr. 4: 78078109 - 78133666 on Build Rnor_6.0
UniGene ID
Rn.124116
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007016.1 NP_001007017.1 17 2661
XM_008762902.1 XP_008761124.1 18 4162
GenBank mRNA AJ629845.1 - 17 2661

View Full Product Details