Gene Transcripts

Gene Symbol
Vom1r49
Entrez Gene ID
494269
Gene Name
vomeronasal 1 receptor 49
Gene Aliases
V1rg8
Chromosome Location
Chr. 1: 66969158 - 66970140 on Build Rnor_6.0
UniGene ID
Rn.129418
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008920.1 NP_001008920.1 583
GenBank mRNA AY510303.1 - 583

View Full Product Details