Gene Transcripts

Gene Symbol
Vom1r96
Entrez Gene ID
494275
Gene Name
vomeronasal 1 receptor 96
Gene Aliases
V1rb14
Chromosome Location
Chr. 4: 121870699 - 121871658 on Build Rnor_6.0
UniGene ID
Rn.129645
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008927.1 NP_001008927.1 1 803
GenBank mRNA AY510296.1 - 1 803

View Full Product Details