Gene Transcripts

Gene Symbol
Shisa3
Entrez Gene ID
498356
Gene Name
shisa family member 3
Gene Aliases
RGD1565710
Chromosome Location
Chr. 14: 42216653 - 42221225 on Build Rnor_6.0
UniGene ID
Rn.198968
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109087.2 NP_001102557.1 1 631
GenBank mRNA BC168960.1 - 1 631

View Full Product Details