Gene Transcripts

Gene Symbol
Pin4
Entrez Gene ID
684441
Gene Name
protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin)
Gene Aliases
Chromosome Location
Chr. X: 72004415 - 72011667 on Build Rnor_6.0
UniGene ID
Rn.160194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752069.3 XP_003752117.1 4 362
XM_006257122.2 XP_006257184.1 4 405
GenBank mRNA FQ222828.1 - 4 305

View Full Product Details