Gene Transcripts

Gene Symbol
Sprr1b
Entrez Gene ID
685912
Gene Name
small proline-rich protein 1B
Gene Aliases
Chromosome Location
Chr. 2: 192623196 - 192623681 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001065728.2 XP_001065728.1 1 410
XM_008761260.1 XP_008759482.1 3 257

View Full Product Details