Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001394.6 | NP_001385.1 | 3 | 1252 | |
NM_057158.3 | NP_476499.1 | 4 | 2067 | ||
XM_011544428.2 | XP_011542730.1 | 3 | 3812 | ||
GenBank mRNA | AK314820.1 | - | 3 | 1177 | |
AL137704.1 | - | 4 | 2067 | ||
BC002671.2 | - | 3 | 938 | 3605895 | |
BC014565.2 | - | 3 | 1170 | 3050199 | |
BG760517.1 | - | 4 | 460 | ||
S80632.1 | - | 3 | 831 | ||
U21108.1 | - | 3 | 1026 | ||
U48807.1 | - | 3 | 899 |