Gene Transcripts

Gene Symbol
TSC2
Entrez Gene ID
7249
Gene Name
tuberous sclerosis 2
Gene Aliases
LAM, PPP1R160, TSC4
Chromosome Location
Chr. 16: 2047804 - 2088720 on Build GRCh38
UniGene ID
Hs.90303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000548.4 NP_000539.2 6 747
NM_001077183.2 NP_001070651.1 6 747
NM_001114382.2 NP_001107854.1 6 747
NM_001318827.1 NP_001305756.1 5 636
NM_001318829.1 NP_001305758.1 5 580
NM_001318832.1 NP_001305761.1 6 648
XM_005255529.4 XP_005255586.2 6 634
XM_005255531.4 XP_005255588.2 6 634
XM_011522636.2 XP_011520938.1 6 634
XM_011522637.2 XP_011520939.1 6 634
XM_011522638.2 XP_011520940.2 5 727
XM_011522639.2 XP_011520941.1 6 634
XM_011522640.2 XP_011520942.1 6 634
XM_017023615.1 XP_016879104.1 6 634
XM_017023616.1 XP_016879105.1 6 634
XM_017023617.1 XP_016879106.1 5 727
XM_017023618.1 XP_016879107.1 6 634
GenBank mRNA AB210000.1 - 6 653
AK295672.1 - 5 467
AK295728.1 - 6 648
BC150300.1 - 6 622 8860381
BF194994.1 - 3 254
KJ534954.1 - 5 547
KJ534955.1 - 5 547
KJ534958.1 - 5 547
KJ534982.1 - 5 547
KJ534984.1 - 5 547
KJ535039.1 - 5 547
KJ535091.1 - 5 547
X75621.1 - 5 565

View Full Product Details