Gene Transcripts
- Gene Symbol
- Pex16
- Entrez Gene ID
- 18633
- Gene Name
- peroxisomal biogenesis factor 16
- Gene Aliases
- Chromosome Location
- Chr. 2: 92374676 - 92381220 on Build GRCm38
- UniGene ID
- Mm.202782
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028114.1 |
|
1 |
95 |
|
GenBank mRNA |
AK020743.1 |
- |
1 |
96 |
|
AK162716.1 |
- |
1 |
86 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY724441.1 |
- |
1 |
89 |
|
Chr. 2 - 92374682 - 92378848 [+] |
View Full Product Details