Gene Transcripts
- Gene Symbol
- 6430411K18Rik
- Entrez Gene ID
- 76880
- Gene Name
- RIKEN cDNA 6430411K18 gen
- Gene Aliases
- PEG11as
- Chromosome Location
- Chr. 12: 109591537 - 109592851 on Build GRCm38
- UniGene ID
- Mm.490311
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002848.3 |
|
1 |
1181 |
|
GenBank mRNA |
AK018276.1 |
- |
1 |
1195 |
|
EU434921.1 |
- |
|
2165 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB590426.1 |
- |
1 |
542 |
|
Chr. 12 - 109592176 - 109592882 [+] |
View Full Product Details