Gene Transcripts
- Gene Symbol
- Zp4-ps
- Entrez Gene ID
- 664793
- Gene Name
- zona pellucida glycoprotein 4, pseudogene
- Gene Aliases
- EG664793, Zp4, Zpb
- Chromosome Location
- Chr. 13: 11522052 - 11525682 on Build GRCm38
- UniGene ID
- Mm.410565
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027813.1 |
|
6 |
781 |
|
GenBank mRNA |
DQ279480.1 |
- |
6 |
781 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV655471.1 |
- |
6 |
679 |
|
Chr. 13 - 11522154 - 11526436 [+] |
View Full Product Details