Gene Transcripts

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
Chromosome Location
Chr. 6: 31173735 - 31177899 on Build GRCh38
UniGene ID
Hs.670091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026816.1 3 454
GenBank mRNA AB932952.1 - 2 421
AB932953.1 - 3 648
AB932954.1 - 3 452
AB933312.1 - 4 719
AB933313.1 - 4 485
AB933320.1 - 6 937
AB933321.1 - 4 633
AY484516.1 - 3 454
LC027944.1 - 3 688
LC027945.1 - 4 746
LC027946.1 - 3 713
LC050981.1 - 5 930
LC050982.1 - 5 1011
LC050983.1 - 5 777
LC050984.1 - 3 485
LC050985.1 - 4 606
LC050986.1 - 6 963
LC050987.1 - 7 1230
LC050988.1 - 2 452
LC050989.1 - 4 744
LC050990.1 - 6 816
LC050991.1 - 4 980

View Full Product Details