Gene Transcripts

Gene Symbol
MRS2P2
Entrez Gene ID
729633
Gene Name
MRS2 pseudogene 2
Gene Aliases
Chromosome Location
Chr. 12: 71848292 - 71850983 on Build GRCh38
UniGene ID
Hs.716256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024072.2 1 1776
GenBank mRNA AY333753.1 - 1777

View Full Product Details