Gene Transcripts
- Gene Symbol
- PART1
- Entrez Gene ID
- 25859
- Gene Name
- prostate androgen-regulated transcript 1 (non-protein coding)
- Gene Aliases
- NCRNA00206
- Chromosome Location
- Chr. 5: 60487713 - 60547657 on Build GRCh38
- UniGene ID
- Hs.146312
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA930113.1 |
- |
1 |
117 |
|
Chr. 5 - 60488140 - 60546164 [+] |
GenBankmRNA |
DA162090.1 |
- |
1 |
74 |
|
Chr. 5 - 60488181 - 60534011 [+] |
GenBankmRNA |
DA296238.1 |
- |
1 |
278 |
|
Chr. 5 - 60487977 - 60529508 [+] |
View Full Product Details