Gene Transcripts

Gene Symbol
PART1
Entrez Gene ID
25859
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
Gene Aliases
NCRNA00206
Chromosome Location
Chr. 5: 60487713 - 60547657 on Build GRCh38
UniGene ID
Hs.146312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024617.1 1 673
NR_028508.1 1 454
NR_028509.1 1 454
GenBank mRNA AF163475.1 - 1 454
AK094518.1 - 1 135
AL050198.1 - 1 197


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA133886.1 - 1 135 Chr. 5 - 60488251 - 60546255 [+]
GenBankmRNA DA930113.1 - 1 248 Chr. 5 - 60488140 - 60546164 [+]
GenBankmRNA DA162090.1 - 1 205 Chr. 5 - 60488181 - 60534011 [+]
GenBankmRNA DA296238.1 - 1 409 Chr. 5 - 60487977 - 60529508 [+]

View Full Product Details