Gene Transcripts

Gene Symbol
OR5E1P
Entrez Gene ID
26343
Gene Name
olfactory receptor family 5 subfamily E member 1 pseudogene
Gene Aliases
HSTPCR24, OR5E1, TPCR24
Chromosome Location
Chr. 11: 7849051 - 7849571 on Build GRCh38
UniGene ID
Hs.382973
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027711.1
1 486
GenBank mRNA AF309699.1 - LIT2079
OR5E1P.
1 486
X89671.1 -
1 463

View Full Product Details