Gene Transcripts

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
Chromosome Location
Chr. 9: 35406755 - 35483029 on Build GRCh38
UniGene ID
Hs.575661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003581.2 3 196
NR_003582.1 3 200
NR_110320.1 3 196
GenBank mRNA AK310278.1 - 3 172
BC031276.1 - 3 183
BC035229.1 - 3 162


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI559666.1 - 3 189 Chr. 9 - 35406772 - 35446491 [+]
GenBankmRNA DB461154.1 - 3 166 Chr. 9 - 35406789 - 35447316 [+]
GenBankmRNA DB456757.1 - 3 165 Chr. 9 - 35406786 - 35444419 [+]
GenBankmRNA DB450766.1 - 3 187 Chr. 9 - 35406764 - 35444405 [+]
GenBankmRNA DB461690.1 - 3 180 Chr. 9 - 35406771 - 35440454 [+]
GenBankmRNA DB459634.1 - 3 180 Chr. 9 - 35406771 - 35440454 [+]
GenBankmRNA DB040474.1 - 3 172 Chr. 9 - 35406779 - 35444828 [+]
GenBankmRNA AA300854.1 - 2 140 Chr. 9 - 35408926 - 35444460 [+]
GenBankmRNA BG720994.1 - 167 Chr. - - [-]
GenBankmRNA DB456494.1 - 3 166 Chr. 9 - 35406789 - 35447278 [+]
GenBankmRNA DB022533.1 - 4 265 Chr. 9 - 35406797 - 35440443 [+]

View Full Product Details