Gene Transcripts

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
Chromosome Location
Chr. 9: 35406755 - 35483029 on Build GRCh38
UniGene ID
Hs.575661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003581.2 5 397
NR_003582.1 5 401
NR_110320.1 5 397
GenBank mRNA AK310278.1 - 5 373
BC031276.1 - 5 384
BC035229.1 - 5 397


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI559666.1 - 5 390 Chr. 9 - 35406772 - 35446491 [+]
GenBankmRNA DB456757.1 - 5 366 Chr. 9 - 35406786 - 35444419 [+]
GenBankmRNA DB450766.1 - 5 388 Chr. 9 - 35406764 - 35444405 [+]
GenBankmRNA DB461690.1 - 5 381 Chr. 9 - 35406771 - 35440454 [+]
GenBankmRNA BI562301.1 - 5 403 Chr. 9 - 35406755 - 35440414 [+]
GenBankmRNA DB459634.1 - 5 381 Chr. 9 - 35406771 - 35440454 [+]
GenBankmRNA DB040474.1 - 5 373 Chr. 9 - 35406779 - 35444828 [+]
GenBankmRNA BG720994.1 - 402 Chr. - - [-]
GenBankmRNA DB022533.1 - 6 466 Chr. 9 - 35406797 - 35440443 [+]

View Full Product Details