Gene Transcripts
- Gene Symbol
- INE1
- Entrez Gene ID
- 8552
- Gene Name
- inactivation escape 1 (non-protein coding)
- Gene Aliases
- NCRNA00010
- Chromosome Location
- Chr. X: 47204848 - 47205861 on Build GRCh38
- UniGene ID
- Hs.657350
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL601447.1 |
- |
1 |
277 |
|
Chr. X - 47205266 - 47206071 [+] |
GenBankmRNA |
DA392789.1 |
- |
1 |
336 |
|
Chr. X - 47205207 - 47205798 [+] |
View Full Product Details