Gene Transcripts
- Gene Symbol
- SPDYE8P
- Entrez Gene ID
- 728524
- Gene Name
- speedy/RINGO cell cycle regulator family member E8, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 73019731 - 73029762 on Build GRCh38
- UniGene ID
- Hs.571275
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003664.2 |
|
2 |
569 |
|
GenBank mRNA |
BC045636.1 |
- |
2 |
572 |
|
BC068610.1 |
- |
2 |
572 |
|
- Gene Symbol
- SPDYE11
- Entrez Gene ID
- 100996746
- Gene Name
- speedy/RINGO cell cycle regulator family member E11
- Gene Aliases
- Chromosome Location
- Chr. 7: 73049256 - 73056645 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- SPDYE17
- Entrez Gene ID
- 102723849
- Gene Name
- speedy/RINGO cell cycle regulator family member E17
- Gene Aliases
- Chromosome Location
- Chr. 7: 77023682 - 77031104 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details