Gene Transcripts

Gene Symbol
TMEM185B
Entrez Gene ID
79134
Gene Name
transmembrane protein 185B
Gene Aliases
FAM11B
Chromosome Location
Chr. 2: 120217471 - 120223408 on Build GRCh38
UniGene ID
Hs.376722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024121.2 NP_077026.2 1 1982
GenBank mRNA AF530474.1 - 1 1982
AK289880.1 - 1 1944


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR743649.1 - 1 220 Chr. 2 - 120221184 - 120221645 [-]
GenBankmRNA DB277964.1 - 1 199 Chr. 2 - 120221057 - 120221624 [-]
GenBankmRNA DA337541.1 - 1 357 Chr. 2 - 120221209 - 120221782 [-]
GenBankmRNA DA221735.1 - 1 64 Chr. 2 - 120220933 - 120221490 [-]
GenBankmRNA DA116246.1 - 1 357 Chr. 2 - 120221205 - 120221783 [-]
GenBankmRNA CN373477.1 - 1 212 Chr. 2 - 120221120 - 120221630 [-]
GenBankmRNA CN373479.1 - 1 43 Chr. 2 - 120220983 - 120221469 [-]

View Full Product Details