Gene Transcripts
- Gene Symbol
- MIR10A
- Entrez Gene ID
- 406902
- Gene Name
- microRNA 10a
- Gene Aliases
- MIRN10A, hsa-mir-10a, miRNA10A, mir-10a
- Chromosome Location
- Chr. 17: 48579838 - 48579947 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029608.1 |
|
1 |
32 |
|
GenBank mRNA |
AJ550394.1 |
- |
|
11 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN286812.1 |
- |
1 |
196 |
|
Chr. 17 - 48579630 - 48580111 [-] |
View Full Product Details