Gene Transcripts
- Gene Symbol
- SNORA5A
- Entrez Gene ID
- 654319
- Gene Name
- small nucleolar RNA, H/ACA box 5A
- Gene Aliases
- ACA5, ACA5A/B/C
- Chromosome Location
- Chr. 7: 45104349 - 45104482 on Build GRCh38
- UniGene ID
- Hs.689711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002919.1 |
|
1 |
17 |
|
GenBank mRNA |
AJ609453.1 |
- |
1 |
17 |
|
View Full Product Details