Gene Transcripts

Gene Symbol
FAM66B
Entrez Gene ID
100128890
Gene Name
family with sequence similarity 66 member B
Gene Aliases
FAM66E
Chromosome Location
Chr. 8: 7301611 - 7355354 on Build GRCh38
UniGene ID
Hs.694680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027423.1 1, 2 550
GenBank mRNA AK302471.1 - 1, 2 550


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DC341951.1 - 1, 2 505 Chr. 8 - 7325177 - 7355309 [-]
GenBankmRNA DC321412.1 - 1, 2 550 Chr. 8 - 7325148 - 7355354 [-]
GenBankmRNA DC348216.1 - 1, 2 541 Chr. 8 - 7349117 - 7355345 [-]
GenBankmRNA DC342600.1 - 1, 2 548 Chr. 8 - 7349129 - 7355352 [-]
GenBankmRNA DC318167.1 - 1, 2 496 Chr. 8 - 7325155 - 7355300 [-]

View Full Product Details