Gene Transcripts
- Gene Symbol
- LRRC27
- Entrez Gene ID
- 80313
- Gene Name
- leucine rich repeat containing 27
- Gene Aliases
- Chromosome Location
- Chr. 10: 132330063 - 132381506 on Build GRCh38
- UniGene ID
- Hs.119897
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB499829.1 |
- |
1 |
36 |
|
Chr. 10 - 132337132 - 132344577 [+] |
GenBankmRNA |
DB493310.1 |
- |
1 |
36 |
|
Chr. 10 - 132337132 - 132344569 [+] |
GenBankmRNA |
DB485255.1 |
- |
1 |
36 |
|
Chr. 10 - 132337132 - 132337698 [+] |
GenBankmRNA |
BX436744.2 |
- |
1 |
114 |
|
Chr. 10 - 132337054 - 132337904 [+] |
GenBankmRNA |
DB505967.1 |
- |
1 |
36 |
|
Chr. 10 - 132337132 - 132344584 [+] |
View Full Product Details