Gene Transcripts
- Gene Symbol
- NRADDP
- Entrez Gene ID
- 100129354
- Gene Name
- neurotrophin receptor associated death domain, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 3: 47011542 - 47013467 on Build GRCh38
- UniGene ID
- Hs.201522
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024046.1 |
|
4 |
563 |
|
GenBank mRNA |
BX117367.1 |
- |
1 |
134 |
|
View Full Product Details