Gene Transcripts

Gene Symbol
NXF5
Entrez Gene ID
55998
Gene Name
nuclear RNA export factor 5
Gene Aliases
Chromosome Location
Chr. X: 101832112 - 101857577 on Build GRCh38
UniGene ID
Hs.307077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028089.1 16, 17 1537
GenBank mRNA AJ277654.1 - 16, 17 1537
AJ277655.1 - 16, 17 1512
AJ277656.1 - 16, 17 1448
AJ277657.1 - 14, 15 1323
AJ277658.1 - 14, 15 1442

View Full Product Details