Gene Transcripts
- Gene Symbol
- POPDC3
- Entrez Gene ID
- 64208
- Gene Name
- popeye domain containing 3
- Gene Aliases
- POP3, bA355M14.1
- Chromosome Location
- Chr. 6: 105157900 - 105179983 on Build GRCh38
- UniGene ID
- Hs.458336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024539.1 |
|
2, 3 |
424 |
|
GenBank mRNA |
AK225989.1 |
- |
2, 3 |
424 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM800745.1 |
- |
2, 3 |
462 |
|
Chr. 6 - 105158372 - 105163765 [-] |
GenBankmRNA |
BU156328.1 |
- |
1, 2 |
219 |
|
Chr. 6 - 105158173 - 105162009 [-] |
GenBankmRNA |
BP369262.1 |
- |
2, 3 |
424 |
|
Chr. 6 - 105158709 - 105176255 [-] |
GenBankmRNA |
CK005752.1 |
- |
2, 3 |
472 |
|
Chr. 6 - 105158642 - 105179940 [-] |
View Full Product Details