Gene Transcripts
- Gene Symbol
- C7orf13
- Entrez Gene ID
- 129790
- Gene Name
- chromosome 7 open reading frame 13
- Gene Aliases
- MY040
- Chromosome Location
- Chr. 7: 156638366 - 156640654 on Build GRCh38
- UniGene ID
- Hs.647014
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA335849.1 |
- |
1 |
519 |
|
Chr. 7 - 156638375 - 156638967 [-] |
GenBankmRNA |
BM713827.1 |
- |
1 |
159 |
|
Chr. 7 - 156638374 - 156638607 [-] |
View Full Product Details