Gene Transcripts
- Gene Symbol
- LRCH3
- Entrez Gene ID
- 84859
- Gene Name
- leucine rich repeats and calponin homology domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 197790855 - 197889346 on Build GRCh38
- UniGene ID
- Hs.518414
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA559512.1 |
- |
1 |
86 |
|
Chr. 3 - 197886457 - 197887271 [+] |
GenBankmRNA |
DA380104.1 |
- |
1 |
24 |
|
Chr. 3 - 197886519 - 197887057 [+] |
View Full Product Details