Gene Transcripts
- Gene Symbol
- XPNPEP1
- Entrez Gene ID
- 7511
- Gene Name
- X-prolyl aminopeptidase 1
- Gene Aliases
- APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
- Chromosome Location
- Chr. 10: 109864766 - 109923553 on Build GRCh38
- UniGene ID
- Hs.390623
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094900.1 |
- |
1 |
1102 |
|
View Full Product Details