Gene Transcripts
- Gene Symbol
- GS1-204I12.4
- Entrez Gene ID
- 105371653
- Gene Name
- uncharacterized GS1-204I12.4
- Gene Aliases
- Chromosome Location
- Chr. 1: 185646373 - 185657168 on Build GRCh38
- UniGene ID
- Hs.738511
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF508908.1 |
- |
2 |
641 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE906931.1 |
- |
3 |
669 |
|
Chr. 1 - 185651167 - 185656997 [+] |
View Full Product Details